Assay Details
Gene Symbol: GFI1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012719.1 | NP_001012737.1 | 2 - 3 | 251 | 153 | |
XM_022419500.1 | XP_022275208.1 | 2 - 3 | 312 | 153 | ||
GenBank mRNA | AB178767.1 | - | 2 - 3 | 281 | 153 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
