Assay Details
Gene Symbol: CCL7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001010960.1 | NP_001010960.1 | 1 - 2 | 76 | 135 | |
GenBank mRNA | AB183286.1 | - | 1 - 2 | 76 | 135 | |
AY849795.1 | - | 1 - 2 | 76 | 135 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
