Assay Details
Gene Symbol: COMT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004074.1 | NP_001004074.1 | 2 - 3 | 411 | 113 | |
XM_005636532.3 | XP_005636589.1 | 3 - 4 | 604 | 113 | ||
XM_005636533.3 | XP_005636590.1 | 3 - 4 | 616 | 113 | ||
XM_014108109.2 | XP_013963584.1 | 3 - 4 | 852 | 113 | ||
GenBank mRNA | AB193089.1 | - | 2 - 3 | 338 | 113 | |
AY340812.1 | - | 2 - 3 | 411 | 113 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
