Assay Details
Gene Symbol: HTR2C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006648.1 | NP_001006649.1 | 3 - 4 | 544 | 85 | |
XM_005640991.3 | XP_005641048.1 | 5 - 6 | 1260 | 85 | ||
GenBank mRNA | AB193091.1 | - | 3 - 4 | 544 | 85 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
