Assay Details
Gene Symbol: F3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024640.1 | NP_001019811.1 | 5 - 6 | 803 | 87 | |
GenBank mRNA | AB200288.1 | - | 5 - 6 | 803 | 87 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
