Assay Details
Gene Symbol: CXCL8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003200.1 | NP_001003200.1 | 2 - 3 | 206 | 65 | |
GenBank mRNA | AF048717.1 | - | 2 - 3 | 246 | 65 | |
D28772.1 | - | 2 - 3 | 235 | 65 | ||
U10308.1 | - | 2 - 3 | 206 | 65 | ||
Z70048.1 | - | 1 - 2 | 137 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
