Assay Details
Gene Symbol: IL4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003159.1 | NP_001003159.1 | 3 - 4 | 367 | 97 | |
GenBank mRNA | AF054833.1 | - | 3 - 4 | 304 | 97 | |
AF083270.1 | - | 3 - 4 | 346 | 97 | ||
AF091132.1 | - | 3 - 4 | 206 | 97 | ||
AF104245.1 | - | 3 - 4 | 304 | 97 | ||
AF187322.1 | - | 3 - 4 | 370 | 97 | ||
AF239917.1 | - | 3 - 4 | 367 | 97 | ||
EF095771.1 | - | 3 - 4 | 314 | 97 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
