Assay Details
Gene Symbol: GH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003168.1 | NP_001003168.1 | 501 | 93 | ||
GenBank mRNA | AF069071.1 | - | 501 | 93 | ||
S78184.1 | - | 337 | 93 | |||
Z23067.1 | - | 447 | 93 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
