Assay Details
Gene Symbol: TIMP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003082.1 | NP_001003082.1 | 343 | 88 | ||
GenBank mRNA | AF095638.1 | - | 201 | 88 | ||
AF112115.1 | - | 343 | 88 | |||
AF188489.1 | - | 138 | 88 |

More Information
Set Membership: |
![]() ![]() |
Gene Ontology Categories:
