Assay Details
Gene Symbol: FLT3LG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003350.1 | NP_001003350.1 | 1 - 2 | 67 | 118 | |
XM_005615738.3 | XP_005615795.1 | 2 - 3 | 227 | 118 | ||
XM_005615739.3 | XP_005615796.1 | 2 - 3 | 170 | 118 | ||
XM_014114041.2 | XP_013969516.1 | 2 - 3 | 192 | 118 | ||
XM_022417411.1 | XP_022273119.1 | 2 - 3 | 188 | 118 | ||
XM_022417428.1 | XP_022273136.1 | 2 - 3 | 193 | 118 | ||
XM_022417456.1 | XP_022273164.1 | 2 - 3 | 117 | 118 | ||
GenBank mRNA | AF155148.1 | - | 1 - 2 | 67 | 118 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001003350.1, XM_005615738.3, XM_005615739.3, XM_014114041.2, XM_022417411.1, XM_022417428.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
