Assay Details
Gene Symbol: IL13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003384.1 | NP_001003384.1 | 2 - 3 | 230 | 72 | |
XM_022424906.1 | XP_022280614.1 | 2 - 3 | 824 | 72 | ||
GenBank mRNA | AF244915.1 | - | 2 - 3 | 230 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
