Assay Details
Gene Symbol: SLCO1B3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001166047.2 | NP_001159519.2 | 6 - 7 | 767 | 141 | |
XM_005636672.3 | XP_005636729.1 | 7 - 8 | 868 | 141 | ||
XM_005636673.3 | XP_005636730.1 | 6 - 7 | 810 | 141 | ||
XM_022410841.1 | XP_022266549.1 | 6 - 7 | 1055 | 141 | ||
GenBank mRNA | AF403244.1 | - | 1 - 2 | 111 | 141 | |
GQ497899.1 | - | 6 - 7 | 767 | 141 | ||
LN833579.1 | - | 6 - 7 | 737 | 141 |

More Information
Set Membership: |
![]() ![]() ![]() |
