Assay Details
Gene Symbol: TNNI3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003041.1 | NP_001003041.1 | 5 - 6 | 421 | 109 | |
GenBank mRNA | AF506750.1 | - | 5 - 6 | 421 | 109 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
