Assay Details
Gene Symbol: ID3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003025.2 | NP_001003025.1 | 1 - 2 | 364 | 76 | |
GenBank mRNA | AJ271644.1 | - | 1 - 2 | 376 | 76 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
