Assay Details
Gene Symbol: CDKN1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005627279.3 | XP_005627336.1 | 2 - 3 | 536 | 64 | |
XM_532125.6 | XP_532125.3 | 2 - 3 | 555 | 64 | ||
GenBank mRNA | AJ830019.1 | - | 1 - 2 | 349 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
