Assay Details
Gene Symbol: TGFA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_022424867.1 | XP_022280575.1 | 3 - 4 | 373 | 67 | |
GenBank mRNA | AY458143.1 | - | 1 - 2 | 68 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
