Assay Details
Gene Symbol: OAS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001048131.1 | NP_001041596.1 | 233 | 53 | ||
XM_005636024.3 | XP_005636081.1 | 287 | 53 | |||
GenBank mRNA | AY863104.1 | - | 233 | 53 |

More Information
Set Membership: |
![]() ![]() ![]() |
