Assay Details
Gene Symbol: HACD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025269.1 | NP_001020440.1 | 318 | 71 | ||
NR_131895.1 | 318 | 71 | ||||
GenBank mRNA | AJ876904.1 | - | 318 | 71 | ||
AJ876905.1 | - | 318 | 71 |

More Information
Set Membership: |
![]() ![]() |
Gene Ontology Categories:
