Assay Details
Gene Symbol: PPP1CA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003064.2 | NP_001003064.2 | 1 - 2 | 152 | 89 | |
GenBank mRNA | AY062037.1 | - | 1 - 2 | 58 | 89 | |
DN365927.1 | - | 1 - 2 | 70 | 89 | ||
DN392813.1 | - | 1 - 2 | 161 | 89 | ||
DN871864.1 | - | 1 - 2 | 69 | 89 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
