Assay Details
Gene Symbol: MITF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003337.1 | NP_001003337.1 | 169 | 88 | ||
XM_005632019.3 | XP_005632076.1 | 52 | 88 | |||
GenBank mRNA | AY240952.1 | - | 169 | 88 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001003337.1, XM_005632019.3 transcripts.
Set Membership: |
![]() ![]() |
