Assay Details
Gene Symbol: MITF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003337.1 | NP_001003337.1 | 386 | 70 | ||
XM_005632016.2 | XP_005632073.1 | 601 | 70 | |||
XM_005632018.2 | XP_005632075.1 | 601 | 70 | |||
XM_005632019.3 | XP_005632076.1 | 269 | 70 | |||
GenBank mRNA | AY240952.1 | - | 386 | 70 |

More Information
Set Membership: |
![]() ![]() |
