Assay Details
Gene Symbol: MITF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003337.1 | NP_001003337.1 | 574 | 81 | ||
XM_005632016.2 | XP_005632073.1 | 789 | 81 | |||
XM_005632018.2 | XP_005632075.1 | 789 | 81 | |||
XM_005632019.3 | XP_005632076.1 | 457 | 81 | |||
GenBank mRNA | AY240952.1 | - | 574 | 81 |
