Assay Details
Gene Symbol: HPRT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003357.2 | NP_001003357.2 | 3 - 4 | 458 | 136 | |
NM_001313818.1 | NP_001300747.1 | 2 - 3 | 351 | 136 | ||
GenBank mRNA | AY283372.1 | - | 3 - 4 | 309 | 136 | |
DN394227.1 | - | 3 - 4 | 468 | 136 | ||
DN418028.1 | - | 2 - 3 | 337 | 136 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
