Assay Details
Gene Symbol: HPRT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003357.2 | NP_001003357.2 | 4 - 5 | 535 | 124 | |
NM_001313818.1 | NP_001300747.1 | 3 - 4 | 428 | 124 | ||
GenBank mRNA | AY283372.1 | - | 4 - 5 | 386 | 124 | |
DN394227.1 | - | 4 - 5 | 545 | 124 | ||
DN418028.1 | - | 3 - 4 | 414 | 124 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
