Assay Details
Gene Symbol: ICOS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002972.2 | NP_001002972.1 | 2 - 3 | 410 | 124 | |
XM_022414940.1 | XP_022270648.1 | 3 - 4 | 671 | 124 | ||
XM_022414941.1 | XP_022270649.1 | 3 - 4 | 554 | 124 | ||
GenBank mRNA | AY342349.1 | - | 2 - 3 | 412 | 124 |

More Information
Set Membership: |
![]() ![]() ![]() |
