Assay Details
Gene Symbol: UNK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003390.1 | NP_001003390.1 | 515 | 60 | ||
XM_022422543.1 | XP_022278251.1 | 1721 | 60 | |||
XM_022422544.1 | XP_022278252.1 | 1042 | 60 | |||
XM_022422545.1 | XP_022278253.1 | 1281 | 60 | |||
XM_022422546.1 | XP_022278254.1 | 3219 | 60 | |||
XM_022422547.1 | XP_022278255.1 | 581 | 60 | |||
XM_022422548.1 | XP_022278256.1 | 1617 | 60 | |||
XM_022422549.1 | XP_022278257.1 | 581 | 60 | |||
XM_022422551.1 | XP_022278259.1 | 578 | 60 | |||
XM_022422552.1 | XP_022278260.1 | 580 | 60 | |||
XM_022422553.1 | XP_022278261.1 | 604 | 60 | |||
XM_022422554.1 | XP_022278262.1 | 1813 | 60 | |||
XM_022422555.1 | XP_022278263.1 | 1788 | 60 | |||
XM_022422556.1 | XP_022278264.1 | 1782 | 60 | |||
XM_022422557.1 | XP_022278265.1 | 1451 | 60 | |||
XM_022422558.1 | XP_022278266.1 | 1545 | 60 | |||
XM_022422559.1 | XP_022278267.1 | 1196 | 60 | |||
GenBank mRNA | AY422569.1 | - | 515 | 60 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_022422554.1, XM_022422555.1, XM_022422556.1, XM_022422557.1, XM_022422558.1, XM_022422559.1 transcripts.
Set Membership: |
![]() ![]() |
Gene Ontology Categories:
