Assay Details
Gene Symbol: UNK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003390.1 | NP_001003390.1 | 1123 | 74 | ||
XM_022422543.1 | XP_022278251.1 | 2329 | 74 | |||
XM_022422544.1 | XP_022278252.1 | 1650 | 74 | |||
XM_022422545.1 | XP_022278253.1 | 1889 | 74 | |||
XM_022422546.1 | XP_022278254.1 | 3827 | 74 | |||
XM_022422547.1 | XP_022278255.1 | 1189 | 74 | |||
XM_022422548.1 | XP_022278256.1 | 2225 | 74 | |||
XM_022422552.1 | XP_022278260.1 | 1188 | 74 | |||
XM_022422553.1 | XP_022278261.1 | 1212 | 74 | |||
XM_022422554.1 | XP_022278262.1 | 2421 | 74 | |||
XM_022422555.1 | XP_022278263.1 | 2396 | 74 | |||
XM_022422556.1 | XP_022278264.1 | 2390 | 74 | |||
XM_022422557.1 | XP_022278265.1 | 2059 | 74 | |||
XM_022422558.1 | XP_022278266.1 | 2153 | 74 | |||
XM_022422559.1 | XP_022278267.1 | 1804 | 74 | |||
GenBank mRNA | AY422569.1 | - | 1123 | 74 |

More Information
Set Membership: |
![]() ![]() |
Gene Ontology Categories:
