Assay Details
Gene Symbol: MGMT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003376.1 | NP_001003376.1 | 3 - 4 | 482 | 54 | |
GenBank mRNA | AY423553.1 | - | 3 - 4 | 482 | 54 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
