Assay Details
Gene Symbol: ESRRA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002936.1 | NP_001002936.1 | 2 - 3 | 441 | 97 | |
XM_005631023.2 | XP_005631080.1 | 3 - 4 | 597 | 97 | ||
XM_022405056.1 | XP_022260764.1 | 3 - 4 | 845 | 97 | ||
GenBank mRNA | AY533243.1 | - | 2 - 3 | 441 | 97 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
