Assay Details
Gene Symbol: HMOX1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001194969.1 | NP_001181898.1 | 2 - 3 | 137 | 87 | |
GenBank mRNA | AY563546.1 | - | 1 - 2 | 74 | 87 | |
DN393611.1 | - | 2 - 3 | 365 | 87 |

More Information
Set Membership: |
![]() ![]() ![]() |
