Assay Details
Gene Symbol: HMOX1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001194969.1 | NP_001181898.1 | 4 - 5 | 737 | 97 | |
GenBank mRNA | AY563546.1 | - | 3 - 4 | 674 | 97 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
