Assay Details
Gene Symbol: WRN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_014120071.2 | XP_013975546.1 | 3347 | 62 | ||
XM_014120072.2 | XP_013975547.1 | 3213 | 62 | |||
XM_014120074.2 | XP_013975549.1 | 2920 | 62 | |||
XM_022404081.1 | XP_022259789.1 | 3197 | 62 | |||
GenBank mRNA | AY705790.1 | - | 448 | 62 |

More Information
Set Membership: |
![]() ![]() ![]() |
