Assay Details
Gene Symbol: TAS1R2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001031819.1 | NP_001026989.1 | 4 - 5 | 1465 | 74 | |
GenBank mRNA | AY916758.1 | - | 4 - 5 | 1465 | 74 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
