Assay Details
Gene Symbol: CCKAR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012646.2 | NP_001012664.1 | 3 - 4 | 629 | 97 | |
GenBank mRNA | AY934643.1 | - | 3 - 4 | 629 | 97 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
