Assay Details
Gene Symbol: ADGRE3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001038666.1 | NP_001033755.1 | 14 - 15 | 1913 | 116 | |
GenBank mRNA | DQ227277.1 | - | 14 - 15 | 1913 | 116 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
