Assay Details
Gene Symbol: RAB7A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003316.1 | NP_001003316.1 | 74 | 147 | ||
XM_005632015.2 | XP_005632072.1 | 293 | 147 | |||
GenBank mRNA | M35522.1 | - | 74 | 147 |

More Information
Set Membership: |
![]() ![]() |
Gene Ontology Categories:
