Assay Details
Gene Symbol: FGF7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003237.1 | NP_001003237.1 | 1 - 2 | 361 | 66 | |
XM_005638175.3 | XP_005638232.1 | 2 - 3 | 686 | 66 | ||
GenBank mRNA | U80800.1 | - | 1 - 2 | 361 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
