Assay Details
Gene Symbol: RAB8A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003152.1 | NP_001003152.1 | 2 - 3 | 193 | 64 | |
GenBank mRNA | X56385.1 | - | 193 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
