Assay Details
Gene Symbol: HCN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_014121847.2 | XP_013977322.1 | 6 - 7 | 1676 | 145 | |
GenBank mRNA | AY686751.1 | - | 1 - 2 | 184 | 145 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
