Assay Details
Gene Symbol: HRAS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001287069.1 | NP_001273998.1 | 2 | 215 | 149 | |
NM_001287070.1 | NP_001273999.1 | 2 | 215 | 149 | ||
GenBank mRNA | U62092.3 | - | 2 | 139 | 149 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
