Assay Details
Gene Symbol: CD5L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001308534.1 | NP_001295463.1 | 3 - 4 | 466 | 105 | |
GenBank mRNA | AB915633.1 | - | 3 - 4 | 380 | 105 | |
DN365189.1 | - | 1 - 2 | 118 | 105 |

More Information
Set Membership: |
![]() ![]() ![]() |
