Assay Details
Gene Symbol: TPPP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005620833.3 | XP_005620890.1 | 2 - 3 | 264 | 88 | |
XM_014114090.2 | XP_013969565.1 | 2 - 3 | 334 | 88 | ||
XM_014114091.2 | XP_013969566.1 | 2 - 3 | 341 | 88 | ||
XM_022419356.1 | XP_022275064.1 | 3 - 4 | 427 | 88 |

More Information
Set Membership: |
![]() ![]() ![]() |
