Assay Details
Gene Symbol: RPL15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001313778.1 | NP_001300707.1 | 3 - 4 | 369 | 84 | |
XM_857783.4 | XP_862876.1 | 3 - 4 | 467 | 84 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
