Assay Details
Gene Symbol: NDUFA11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_533939.6 | XP_533939.2 | 1 - 2 | 182 | 83 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
