Assay Details
Gene Symbol: OAZ1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001127234.1 | NP_001120706.1 | 3 - 4 | 620 | 89 | |
GenBank mRNA | DN409262.1 | - | 1 - 2 | 62 | 89 | |
DN430722.1 | - | 620 | 89 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
