Assay Details
Gene Symbol: LOC608697 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_014106509.2 | XP_013961984.1 | 2 - 3 | 240 | 68 | |
XM_014106515.2 | XP_013961990.1 | 2 - 3 | 240 | 68 | ||
XM_022408603.1 | XP_022264311.1 | 2 - 3 | 240 | 68 | ||
XM_022408604.1 | XP_022264312.1 | 2 - 3 | 240 | 68 | ||
XM_022408605.1 | XP_022264313.1 | 1 - 2 | 223 | 68 | ||
XM_022408608.1 | XP_022264316.1 | 1 - 2 | 224 | 68 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_022408605.1, XM_022408608.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
