Assay Details
Gene Symbol: EIF6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_014107003.2 | XP_013962478.1 | 2 - 3 | 380 | 56 | |
XM_014107004.2 | XP_013962479.1 | 2 - 3 | 478 | 56 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
