Assay Details
Gene Symbol: MRPL40 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_534762.6 | XP_534762.3 | 2 - 3 | 252 | 93 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
