Assay Details
Gene Symbol: TASOR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005617174.3 | XP_005617231.1 | 1176 | 120 | ||
XM_005617175.3 | XP_005617232.1 | 1151 | 120 | |||
XM_022408084.1 | XP_022263792.1 | 1151 | 120 | |||
XM_022408087.1 | XP_022263795.1 | 1013 | 120 | |||
XM_022408090.1 | XP_022263798.1 | 1151 | 120 | |||
XM_022408091.1 | XP_022263799.1 | 1151 | 120 |

More Information
Set Membership: |
![]() ![]() |
