Assay Details
Gene Symbol: AK6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001286232.1 | NP_001273161.1 | 2 - 3 | 130 | 154 | |
GenBank mRNA | DN392943.1 | - | 2 - 3 | 130 | 154 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
